A17671.14
A17671.14EA
47.9
EUR
InStock
A17671.14
Chlorure de nonanoyle ≥97%
Chlorure de nonanoyle
Purity: 97%
CAS No.: 764-85-2
Molecular Formula: CH3(CH2)7COCl
Formula Weight: 176.69
Flash Point: 95°(203°F)
Refractive Index: 1.4380
Melting Point: -61°
Boiling Point: 215°
Beilstein Reference: 1362621
UN No.: 3265
MDL No.: MFCD00000768