Synonyms:
Dithizone,for analysis, 1,5-Diphenylthiocarbazone
A17947.09
A17947.09EA
57.4
EUR
InStock
A17947.09
Dithizone ≥98%
Dithizone
Purity: 98%
CAS No.: 60-10-6
Molecular Formula: C13H12N4S
Formula Weight: 256.33
Odour: Odourless
Melting Point: ca 168° dec.
Beilstein Reference: 748838
Merck Reference: 14,3377
MDL No.: MFCD00003025