A18052.06
A18052.06EA
30
EUR
InStock
A18052.06
1,4-Thioxane ≥98%
1,4-Thioxane
Purity: 98+%
CAS No.: 15980-15-1
Molecular Formula: C4H8OS
Formula Weight: 104.17
Flash Point: 42°(107°F)
Refractive Index: 1.5070
Odour: Unpleasant
Boiling Point: 147-148°
Beilstein Reference: 102552
UN No.: 1993
MDL No.: MFCD00006573