Synonyms:
1,3-Di(dimethylethyl)-4-hydroxybenzene
A18509.30
A18509.30EA
30.9
EUR
InStock
A18509.30
2,4-Di-tert-butylphénol 97%
2,4-Di-tert-butylphénol
Purity: 97%
CAS No.: 96-76-4
Molecular Formula: C14H22O
Formula Weight: 206.33
Flash Point: 129°(264°F)
Melting Point: 53-57°
Boiling Point: 265°
Beilstein Reference: 1910383
UN No.: 3077
MDL No.: MFCD00008828