Synonyms:
2,6-Dibromo-4-methylphenol, 3,5-Dibromo-4-hydroxytoluene, 1,3-Dibromo-2-hydroxy-5-methylbenzene
A18634.14
A18634.14EA
53.3
EUR
InStock
A18634.14
2,6-Dibromo-p-crésol ≥98%
2,6-Dibromo-p-crésol
Purity: 98%
CAS No.: 2432-14-6
Molecular Formula: C7H6Br2O
Formula Weight: 265.94
Flash Point: 110°(230°F)
Melting Point: 49-50°
Beilstein Reference: 2045283
MDL No.: MFCD00002154