Synonyms:
Acetic Acid Prenyl Ester, Acetic Acid 3-Methyl-2-butenyl Ester, 3-Methylbut-2-en-1-yl acetate, Prenyl Acetate
Stabilization:
Stabilised with α-tocopherol 0.1%
A18958.18
A18958.18EA
35.4
EUR
InStock
A18958.18
3-Méthyl-2-butén-1-yle acétate 98% stabilisé
3-Méthyl-2-butén-1-yle acétate
Stab. with 0.1% alpha tocopherol
Purity: 98%
CAS No.: 1191-16-8
Molecular Formula: (CH3)2C=CHCH2OCOCH3
Formula Weight: 128.17
Flash Point: 40°(104°F)
Refractive Index: 1.43
Odour: Fruit-like
Boiling Point: 140°
UN No.: UN3272
MDL No.: MFCD00036569