Organic reference standard, alpha-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20), Supelco®
Fournisseur: Merck
A4582-.5ML
A4582-.5MLEA 29.8 EUR InStock
A4582-.5ML
Organic reference standard, alpha-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20), Supelco®
Étalons Étalons organiques
α-Amylase belongs to glycosyl hydrolase family 13.
  • α-Amylase from Bacillus licheniformis
  • Used in two-step hydrolysis for enzymatic saccharification of dried biomass powder
  • Utilised to analyse the starch concentration by an enzymatic method
α-Amylase, the digestive enzyme in saliva has several significance in industries like, food, pharmaceutical, textile and paper. It hydrolyses the linear chain of amylose to form glucose molecules.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité