Synonyms:
1-(3,7-Dihydroxy-10H-phenoxazin-10-yl)ethanone, ADHP, Amplex® Red (Registered trademark of LifeTechnologies)
11000
AATB11000EA
307
EUR
InStock
AATB11000
10-Acetyl-3,7-dihydroxyphenoxazine ≥95%
10-Acetyl-3,7-dihydroxyphenoxazine
Our Amplite® ADHP is chemically same to Amplex™ Red (Amplex™ Red is the trademark of Invitrogen).