Amplite® Blue Peroxidase Substrate
Fournisseur: AAT BIOQUEST INC
11005
AATB11005EA 221 EUR InStock
AATB11005
Amplite® Blue Peroxidase Substrate
Tests Tests enzymatiques
A variety of substrates are available for detecting peroxidase activity in ELISA assays. colourimetric HRP substrates (e.g., TMB, OPD and ABTS) have been widely used for years. Each of these substrates varies greatly with respect to its performance characteristics such as detection sensitivity, working range and attainable signal-to-noise ratios. Amplite® Blue peroxidase substrate is a soluble fluorogenic substrate for detection of peroxidase activity. Amplite® Blue allows rapid HRP detection assays to be performed with greater sensitivity than the colourimetric HRP substrates (such as TMB and OPD). The fluorescent product of Amplite® Blue does not photobleach. Amplite® Blue exhibits a flat baseline in assays, which facilitates low-level detection sensitivity and allows for high signal-to-noise ratios.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité