11061
AATB11061EA
460
EUR
InStock
AATB11061
Azido-Cy5 tyramide
Azido-Cy5 tyramide
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
- Bright signal
- Excellent signal amplification for low expression targets
- Click chemistry building block