Portelite™ Fluorimetric Protein Quantitation Kit
Fournisseur: AAT BIOQUEST INC
11109
AATB11109EA 115 EUR InStock
AATB11109
Portelite™ Fluorimetric Protein Quantitation Kit
Tests Essais protéiniques
Optimised for CytoCite™ and Qubit™ fluorometers. The Portelite™ fluorimetric protein quantitation kit is significantly more sensitive than existing colorimetric protein measurements, e.g., Bradford and Bicinchoninic acid (BCA) assays.
  • Prolite™ Orange used in the kit is non-fluorescent in aqueous solution, but reacts rapidly with proteins and generates bright fluorescence
  • The assay has dynamic range from 12,5 µg/ml to 5 mg/ml of BSA
  • Six-month shelf life
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité