Amplite® Colorimetric Lipase Activity Assay Kit
Fournisseur: AAT BIOQUEST INC
11315
AATB11315EA 647 EUR InStock
AATB11315
Amplite® Colorimetric Lipase Activity Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® colorimetric lipase activity assay kit offers a straightforward and efficient protocol for quantifying lipase activity.
  • Robust protocol
  • Suitable for detecting lipase activity in a variety of biological samples, including cells, supernatant, tissue extracts, and serum
This kit utilises a coupled enzymatic reaction, which converts the substrate into a colored product (586 nm) proportional to the lipase activity present.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité