Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Fournisseur: AAT BIOQUEST INC
11321
AATB11321EA 386 EUR InStock
AATB11321
Amplite® Colorimetric Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® colorimetric GAPDH activity assay kit is a sensitive and simple tool for monitoring GAPDH activity based on a coupled enzymatic reaction catalysed by GAPDH.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six month shelf life
One unit (U) is the amount of enzyme that catalyses the reaction of 1 µmol of substrate per minute.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité