Amplite® Fluorimetric Lipase Activity Assay Kit
Fournisseur: AAT BIOQUEST INC
11322
AATB11322EA 647 EUR InStock
AATB11322
Amplite® Fluorimetric Lipase Activity Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® Fluorimetric Lipase Activity Assay Kit provides a simple and quick protocol for measuring lipase activity.
  • This kit is suitable for detecting lipases from various biological samples like cells, supernatants, tissue extracts, or serum, and is adaptable for high-throughput screening applications
  • Ex/Em=540/610 nm
Lipases belong to a class of enzymes that catalyse the hydrolysis of ester bonds in lipids, such as triglycerides, phospholipids and cholesterol esters.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité