Amplite® Fluorimetric DPP4 Inhibitor Screening Kit
Fournisseur: AAT BIOQUEST INC
11324
AATB11324EA 647 EUR InStock
AATB11324
Amplite® Fluorimetric DPP4 Inhibitor Screening Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® Fluorimetric DPP4 Inhibitor Screening Kit provides an efficient and direct method for evaluating potential DPP4 inhibitors.
  • Ex/Em=360/460 nm
  • Assay is optimised for high-throughput screening systems
This kit includes Sitagliptin, a well-characterised DPP4 inhibitor used in diabetes treatment, as a reference control, facilitating the assessment of inhibitor efficacy.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité