Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Fournisseur: AAT BIOQUEST INC
11327
AATB11327EA 517 EUR InStock
AATB11327
Amplite® Fluorimetric Neutrophil Elastase Activity Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® fluorimetric neutrophil elastase activity assay kit provides a robust and accurate method for quantifying neutrophil elastase (NE) activity.
  • Ex/Em= 360/470 nm
  • Consistent results
  • Six-month shelf life
The kit’s straightforward protocol is optimised for the sensitive detection of NE in blood and plasma samples and is suitable for screening and characterising chemotactic agents triggering neutrophil stimulation.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité