1136
AATB1136EA
295
EUR
InStock
AATB1136
UV460 succinimidyl ester, mFluor™ sonde fluorescente
UV460 succinimidyl ester
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
- pH-insensitive
- High photostability
- Brighter signal