11405
AATB11405EA
146
EUR
InStock
AATB11405
SunRed™ Acetate
SunRed™ Acetate
Esterase-catalyzed hydrolysis of Sun Red acetate (SRA) yields the Sun Red fluorophore that can be excited with the 633 nm laser with emission of ~660 nm.
- Optimised for Cy5 filter set
- Extremely sensitive NIR esterase substrate
- Excellent cell viability stain
- Useful in combination with green vital stains