11622
AATB11622EA
152
EUR
InStock
AATB11622
phosphatase reaction stopping solution, Signal Guard™
phosphatase reaction stopping solution
Alkaline and acid phosphatases are the hydrolase enzymes responsible for dephosphorylation of molecules such as nucleotides, proteins and alkaloids under alkaline and acidic conditions, respectively.
- Compatible with all major phosphatase groups
- Neutral and mild
- Ready to use
- Stabilizes signal for up to 18 hours
- Compatible with colorimetric and fluorimetric substrates