11627
AATB11627EA
146
EUR
InStock
AATB11627
DiFMUP alkaline phosphatase substrate
DiFMUP
DiFMUP is a fluorinated MUP, which is a commonly used fluorogenic alkaline phosphatase substrate.
- Excellent upgrade from MUP
- Useful at low pH
- Multiple applications