Amplite® Colorimetric Gama-Glutamyltransferase (GGT) Activity Assay Kit
Fournisseur: AAT BIOQUEST INC
11801
AATB11801EA 647 EUR InStock
AATB11801
Amplite® Colorimetric Gama-Glutamyltransferase (GGT) Activity Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® Colorimetric Gamma-Glutamyltransferase (GGT) Activity Assay Kit provides an efficient and straightforward method for measuring GGT activity across various sample types.
  • Robust protocol
  • Absorbance measured at 418 nm
  • Six-month shelf life
It is compatible with a wide range of biological samples, including cells, tissue extracts, and serum, and is suitable for high-throughput systems.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité