Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Fournisseur: AAT BIOQUEST INC
11802
AATB11802EA 778 EUR InStock
AATB11802
Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Screen Quest™ fluorimetric neutrophil elastase inhibitor screening kit offers a fast, sensitive, and high-throughput method for screening and characterising potential inhibitors of NE.
  • Ex/Em=360/470nm
  • Consistent results
  • Six-month shelf life
The amount of fluorescent product is directly proportional to the enzymatic activity of the neutrophil elastase present in the sample.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité