12609
AATB12609EA
152
EUR
InStock
AATB12609
NTA maleimide pour la fixation biomoléculaire thiol-surface
NTA maleimide
NTA maleimide is used to attach NTA groups to a biological substrate or a surface that has thiol groups.
- Thiol-reactive
- HIS tag building block
- Multiple applications