13201
AATB13201EA
146
EUR
InStock
AATB13201
D-VLK-AMC [D-Val-Leu-Lys-AMC]
D-VLK-AMC [D-Val-Leu-Lys-AMC]
D-VLK-AMC is a sensitive, highly specific fluorescent substrate for plasmin.
- Bright signal
- High sensitivity
- Suitable for all fluorescent instrumentation