13202
AATB13202EA
137
EUR
InStock
AATB13202
Beta-Ala-AMC
Beta-Ala-AMC
Beta-Ala-AMC is a fluorogenic substrate that might be used for detection pancreatic elastase, arylamidase and beta-alanine aminopeptidase activities.
- Adaptable fluorogenic substrate
- Detects enzymes in multiple species
- Suitable for cell, food, and other sample types