13203
AATB13203EA
292
EUR
InStock
AATB13203
Beta-Ala-R110
Beta-Ala-R110
Beta-Alanine-R110 is a fluorogenic substrate that might be used for detection pancreatic elastase, arylamidase and beta-alanine aminopeptidase activities.
- Adaptable fluorogenic substrate
- Detects enzymes in multiple species
- Suitable for cell, food, and other sample types