13421
AATB13421EA
230
EUR
InStock
AATB13421
Z-DEVD-AMC
Z-DEVD-AMC
Z-DEVD-AMC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
- Caspase detection
- Bright signal