13422
AATB13422EA
152
EUR
InStock
AATB13422
Z-DEVD-pNA
Z-DEVD-pNA
Z-DEVD-pNA is a colorimetric substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.
- Caspase detection
- Highly coloured signal