Synonyms:
H-D-Ala-AMC·TFA, (R)-2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)propanamide 2,2,2-trifluoroacetate
13459
AATB13459EA
152
EUR
InStock
AATB13459
H-D-Ala-AMC·TFA ≥90%
H-D-Ala-AMC·TFA
This D-Ala-AMC is a fluorogenic aminopeptidase and endopeptidase substrate used for the detection of enzymatic activity of microorganisms collected from the surface of aerobically cold-stored pork and beef.