13479
AATB13479EA
146
EUR
InStock
AATB13479
Ac-Pro-Ala-Leu-AMC fluorogenic substrate
Ac-Pro-Ala-Leu-AMC
The weakly fluorescent AMC substrates generate the bright blue fluorescent AMC product that has Ex/Em = 351/430 nm, and can be easily detected with a DAPI filter set.
- High fluorescence
- Rapid activation