13557
AATB13557EA
218
EUR
InStock
AATB13557
Mca-YVADAPK(Dnp)-OH
Mca-YVADAPK(Dnp)-OH
Mca-YVADAPK(Dnp)-OH is a fluorogenic peptide substrate for detecting caspase-1/interleukin-converting enzyme (ICE) and angiotensin I-converting enzyme-2 (ACE-2).
- Caspase detection
- Bright signal