13560
AATB13560EA
209
EUR
InStock
AATB13560
Mca-PLAQAV-Dpa-RSSSR-NH2
Mca-PLAQAV-Dpa-RSSSR-NH2
Mca-PLAQAV-Dpa-RSSSR-NH2 is a fluorogenic peptide substrate for detecting tumor necrosis factor-alpha converting enzyme (TACE or ADAM17) and related enzymes such as ADAM8, ADAM9 and ADAM10.
- Tumor necrosis factor-alpha converting enzyme detection
- Bright signal