13602
AATB13602EA
460
EUR
InStock
AATB13602
FAM-cAMP PDE IV substrat "Green Fluorescence"
FAM-cAMP PDE IV
This green fluorescent cAMP derivative is a specific substrate for phosphodiesterase (PDE) IV.
- Excellent and specific fluorogenic PDE substrate
- cAMP research tool
- Multiple applications