13603
AATB13603EA
460
EUR
InStock
AATB13603
TAMRA-cAMP PDE IV substrat "Red Fluorescence"
TAMRA-cAMP PDE IV
This red cAMP derivative is a specific substrate for phosphodiesterase (PDE) IV.
- Excellent and specific fluorogenic PDE substrate
- cAMP research tool
- Multiple applications