1433
AATB1433EA
769
EUR
InStock
AATB1433
Eu succinimidyl ester, trFluor™ sonde fluorescente
Eu succinimidyl ester
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- No crosslinking
- High stability and selectivity
- Insensitive to fluorescence quenching
- High emission yield