1434
AATB1434EA
460
EUR
InStock
AATB1434
Eu maleimide, trFluor™ sonde fluorescente
Eu maleimide
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- no crosslinking
- High stability and selectivity
- Insensitive to fluorescence quenching
- High emission yield