1500
AATB1500EA
383
EUR
InStock
AATB1500
647 succinimidyl ester, zFluor™ sonde fluorescente
647 succinimidyl ester
Our zFluor™ serial dyes are developed to have the possibly highest photostability at a given wavelength compared to the other similar wavelength dyes on the market.
- High photostability
- Excellent thermal stability
- Bright signal
- pH independence