1510
AATB1510EA
383
EUR
InStock
AATB1510
633 succinimidyl ester, zFluor™ sonde fluorescente
633 succinimidyl ester
This zFluor™ serial dyes are developed to have the possibly highest photostability at a given wavelength compared to the other similar wavelength dyes.
- High photostability
- Excellent thermal stability
- Bright signal
- pH independence