16310
AATB16310EA
614
EUR
InStock
AATB16310
DAX-J2™ Ratio 580/460
DAX-J2™
DAX-J2™ Ratio 580/460 is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.
- Does not require esterase activity for no detection
- Ph-independent fluorescence
- More photostable than daf-2 product
- More sensitive for detecting no than daf-2
- Can be monitored in a ratiometric mode
Attention: For research use only.