trFluor™ Tb-streptavidin conjugate
Fournisseur: AAT BIOQUEST
16926
AATB16926EA 464 EUR InStock
AATB16926
trFluor™ Tb-streptavidin conjugate
trFluor™ Tb-streptavidin
trFluor™ Tb probes are insensitive to fluorescence quenching when conjugated to biological polymers such as antibodies.
  • Stable for 12 months
  • Designed to enable time-resolved fluorometry (TRF)
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
Attention: For Research Use Only
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité