17010
AATB17010EA
460
EUR
InStock
AATB17010
TF5-dUTP 1 mM in TRIS buffer (pH 7.5)
TF5-dUTP
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- Useful for fish probe development
- Well-cited
- Multiple applications
Attention: For research use only.