17029
AATB17029EA
137
EUR
InStock
AATB17029
iFluor® 440-dUTP 1 mM in TRIS buffer (pH 7.5)
iFluor® 440-dUTP
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- Useful for DNA probe development
- Well-cited
- Multiple applications
Attention: For research use only.