17040
AATB17040EA
281
EUR
InStock
AATB17040
XFD™488-dUTP 1 mM in TRIS buffer (pH 7.5)
XFD™488-dUTP
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling, or PCR.
- Useful for DNA probe development
- Well-cited
- Multiple applications
Attention: For research use only.