Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Fournisseur: AAT BIOQUEST INC
17623
AATB17623EA 270 EUR InStock
AATB17623
Helixyte™ Green Fluorimetric ssDNA Quantitation Kit
Tests Tests à l'acide nucléique
Helixyte™ Green fluorimetric ssDNA quantitation kit is optimised for quantifying dsDNA with a fluorescence microplate reader.
  • Optimized for microplate readers
  • High sensitivity
  • Low interference from common contaminants
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution. It enables researchers to quantify as little as 100 pg/ml oligonucleotide or ssDNA. This sensitivity exceeds that achieved with absorbance methods by more than 10000-fold.
With this kit, as little as 1 ng/ml oligonucleotide or ssDNA might be detected.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité