Portelite™ Fluorimetric ssDNA Quantitation Kit
Fournisseur: AAT BIOQUEST
17625
AATB17625EA 409 EUR InStock
AATB17625
Portelite™ Fluorimetric ssDNA Quantitation Kit
Tests Tests à l'acide nucléique
The assay detects long oligonucleotides or ssDNA. Nucleotides and short oligo nucleotides of six bases or less do not interfere in the quantitation assay.
  • Optimized for cytocite and qubit fluorometers
  • Any volume from 1 to 20 µl is acceptable
  • Assay range of 1 to 200 ng
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité