StrandBrite™ Green Fluorimetric RNA Quantitation Kit *Optimised for Microplate Readers*
Fournisseur: AAT BIOQUEST INC
17655
AATB17655EA 442 EUR InStock
AATB17655
StrandBrite™ Green Fluorimetric RNA Quantitation Kit *Optimised for Microplate Readers*
Tests Tests à l'acide nucléique
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité