2025
AATB2025EA
307
EUR
InStock
AATB2025
DNP amine FRET quencher precursor
DNP amine
A carbonyl-reactive building block for developing a probe that can be recognized by anti-DNP antibodies; Also an excellent carbonyl-reactive FRET quencher paired with Trp or Tyr.