2026
AATB2026EA
307
EUR
InStock
AATB2026
DNP maleimide FRET quencher precursor
DNP maleimide
A thiol-reactive building block for developing a probe that can be recognized by anti-DNP antibodies; Also an excellent thiol-reactive FRET quencher paired with Trp or Tyr.