Amplite® Fluorimetric cADP-Ribose Assay Kit
Fournisseur: AAT BIOQUEST INC
20305
AATB20305EA 739 EUR InStock
AATB20305
Amplite® Fluorimetric cADP-Ribose Assay Kit
Tests Tests enzymatiques
This cADP-ribose assay can be performed in a convenient 96-well or 384-well microtiter plate.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
cADP-ribose (cADPR) is a novel Ca2+ messenger derived from NAD+. ADP-riboxyl cyclase (ADPRC) catalyses the synthesis of cADPR from NAD+, but the reaction can be reversed in the presence of high concentration of nicotinamide, producing NAD+ from cADPR stoichiometrically.
The resultant NAD+ can be detected using our newly developed NAD sensor Quest Fluor™ NAD reagent. This makes monitoring cADPR in tissues and cell cultures possible in the low nM range.
The NAD+ detection using Quest Fluor™ NAD reagent is specific to NAD+ and has no reaction to NADH. The fluorescent signal can be readily detected.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité