20506
AATB20506EA
295
EUR
InStock
AATB20506
OG488 BAPTA-1 hexapotassium salt Cell impermeant [equivalent to Oregon Green® 488 BAPTA-1, hexapotassium salt]
OG488 BAPTA-1 hexapotassium salt
OG488 BAPTA -1 is the same molecule of Oregon Green 488 BAPTA-1.
- Calcium-dependent fluorescence
- Steady signal
- Cell impermeant
Attention: For research use only.